siRNA Details
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081229.1 | NP_001074698.1 | 4 | 3350 | ||
GenBank mRNA | AK007760.1 | - | 3 | 1692 | ||
AK156074.1 | - | 2 | 1335 | |||
BC058221.1 | - | 3 | 1545 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191960, XM_006232386.
Gene Ontology Categories:
