siRNA Details
Gene Symbol: Zfp661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111029.1 | NP_001104499.1 | 5 | 1907 | ||
NM_028141.3 | NP_082417.3 | 6 | 2124 | |||
GenBank mRNA | AK013000.1 | - | 1 | 1019 | ||
AK033122.1 | - | 5 | 1909 | |||
AK134651.1 | - | 6 | 3178 | |||
BC052876.1 | - | 6 | 2128 | 30047253 |
