siRNA Details
Gene Symbol: UQCRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199975.2 | NP_001186904.1 | 4, 5 | 484 | ||
NM_006294.4 | NP_006285.1 | 3, 4 | 370 | |||
GenBank mRNA | AK307085.1 | - | 4, 5 | 393 | ||
AK311756.1 | - | 3, 4 | 295 | |||
BC005230.1 | - | 3, 4 | 319 | 3961169 | ||
BI547559.1 | - | 3, 4 | 371 | |||
CR542176.1 | - | 2, 3 | 266 | |||
CR542196.1 | - | 3, 4 | 266 | |||
M22348.1 | - | 3, 4 | 298 | |||
M26700.1 | - | 3, 4 | 318 | |||
X13585.1 | - | 3, 4 | 298 |
Gene Symbol: UQCRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002308.1 | 1 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
