siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177357.3 | NP_796331.2 | 46 | 6524 | ||
GenBank mRNA | AK008844.1 | - | 1 | 102 | ||
AK088732.1 | - | 12 | 1471 | |||
AK139581.1 | - | 13 | 2628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
