siRNA Details
Gene Symbol: Rbm12b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028226.2 | NP_082502.2 | 3 | 1985 | ||
GenBank mRNA | AK013872.1 | - | 1 | 1080 | ||
AK030861.1 | - | 4 | 1693 | |||
BC050844.1 | - | 3 | 1786 | 6513134 | ||
BC054081.1 | - | 1 | 718 |
Gene Symbol: Rbm12b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198957.2 | NP_945195.1 | 4 | 1715 | ||
GenBank mRNA | AK080943.1 | - | 4 | 1645 | ||
AK169895.1 | - | 2 | 1139 | |||
BC030408.1 | - | 1 | 52 | |||
BC080741.1 | - | 3 | 1630 | 30461213 | ||
BC137620.1 | - | 3 | 1730 | 8860639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
