siRNA Details
Gene Symbol: Fgfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028263.1 | NP_082539.2 | 2 | 939 | ||
GenBank mRNA | AK011996.1 | - | 1 | 749 | ||
AK029452.1 | - | 1 | 742 | |||
AK032142.1 | - | 1 | 37 | |||
AK167949.1 | - | 2 | 957 | |||
BC055778.1 | - | 2 | 939 | 6403098 |
