siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028385.1 | NP_082661.1 | 7 | 856 | ||
GenBank mRNA | AK013649.1 | - | 3 | 786 | ||
AK122551.1 | - | 7 | 856 | |||
AK164036.1 | - | 2 | 2761 | |||
BC056502.1 | - | 9 | 1392 | |||
BC083184.1 | - | 6 | 737 | 30536831 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106614, XM_006237014, XM_008763187, XM_006237012, XM_008763189, and others.
Gene Ontology Categories:
