siRNA Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 1, 2 | 304 | ||
GenBank mRNA | AF536771.1 | - | 1, 2 | 73 | ||
AK013600.1 | - | 1, 2 | 306 | |||
AK148956.1 | - | 1, 2 | 275 | |||
BC026686.1 | - | 1, 2 | 242 | 5065516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
