siRNA Details
Gene Symbol: 1700040L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028491.1 | NP_082767.1 | 4, 5 | 541 | ||
GenBank mRNA | AK006661.1 | - | 4, 5 | 542 | ||
AK029622.1 | - | 4, 5 | 541 | |||
BC087900.1 | - | 4, 5 | 531 | 6774064 |

More Information
Other Species Targeted

Gene Ontology Categories:
