siRNA Details
Gene Symbol: 1700052K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006771.1 | - | 1 | 940 | ||
AK148336.1 | - | 1 | 817 | |||
BC099431.1 | - | 1 | 624 | 30525202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_028539.1 | - | 940 | Chr. - - [-] on Build |
