siRNA Details
Gene Symbol: LOC100043315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106998.1 | - | 3 | 1021 | ||
BC106999.1 | - | 3 | 930 | |||
BC114343.1 | - | 3 | 930 | |||
BC115945.1 | - | 3 | 930 | |||
BC115946.1 | - | 3 | 930 | |||
BC127134.1 | - | 3 | 930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
