siRNA Details
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046888.1 | - | 2269 | |||
BC051923.1 | - | 2229 | ||||
BC060063.1 | - | 2229 | ||||
BC061463.1 | - | 2225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
