siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028610.2 | NP_082886.2 | 3 | 337 | ||
GenBank mRNA | AF490348.1 | - | 3 | 254 | ||
AK010753.1 | - | 3 | 305 | |||
AK033197.1 | - | 3 | 338 | |||
BC064754.1 | - | 3 | 276 | 30024281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
