siRNA Details
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028632.2 | NP_082908.2 | 4 | 237 | ||
GenBank mRNA | AK003561.1 | - | 3 | 196 | ||
BC033045.1 | - | 3 | 215 | 1068150 | ||
BC094619.1 | - | 4 | 224 | |||
CJ085020.1 | - | 4 | 252 |
