siRNA Details
Gene Symbol: Haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028633.3 | NP_082909.1 | 6 | 619 | ||
GenBank mRNA | AK003871.1 | - | 3 | 242 | ||
AY029337.1 | - | 6 | 619 | |||
BC025464.1 | - | 6 | 610 | 5351961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
