siRNA Details
Gene Symbol: Ddias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080995.1 | NP_001074464.1 | 5 | 508 | ||
GenBank mRNA | AK014608.2 | - | 4 | 428 | ||
AK154915.1 | - | 4 | 411 | |||
BC066148.1 | - | 4 | 402 | 30086532 | ||
DQ400346.1 | - | 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
