siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177615.1 | NP_001171086.1 | 5 | 829 | ||
NM_001177620.1 | NP_001171091.1 | 7 | 776 | |||
NM_028755.3 | NP_083031.1 | 6 | 738 | |||
GenBank mRNA | AK002950.2 | - | 6 | 722 | ||
AK159004.1 | - | 5 | 808 | |||
AK160860.1 | - | 7 | 774 | |||
BC027107.1 | - | 6 | 726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
