siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 40 | 5259 | ||
GenBank mRNA | AK004816.2 | - | 4 | 397 | ||
AK028968.1 | - | 3 | 317 | |||
AK131180.1 | - | 17 | 2222 | |||
BC030316.1 | - | 17 | 2250 | |||
BC043470.1 | - | 9 | 1062 | |||
BC055295.1 | - | 9 | 1090 | |||
BC141358.1 | - | 40 | 5254 | 9055970 |

More Information
Other Species Targeted

Gene Ontology Categories:
