siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 44, 45 | 5947 | ||
GenBank mRNA | AK004816.2 | - | 8, 9 | 1085 | ||
AK028968.1 | - | 7, 8 | 1005 | |||
AK131180.1 | - | 21, 22 | 2910 | |||
BC029018.1 | - | 3, 4 | 356 | |||
BC030316.1 | - | 21, 22 | 2938 | |||
BC043470.1 | - | 13, 14 | 1750 | |||
BC055295.1 | - | 13, 14 | 1778 | |||
BC141358.1 | - | 44, 45 | 5942 | 9055970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
