siRNA Details
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 20, 21 | 5583 | ||
GenBank mRNA | AK012129.1 | - | 3, 4 | 554 | ||
AK044193.1 | - | 7, 8 | 1410 | |||
AK087819.1 | - | 7, 8 | 1568 | |||
AK172968.1 | - | 14, 15 | 2422 | |||
BC034106.1 | - | 3, 4 | 407 | |||
BC095969.1 | - | 10, 11 | 2113 | |||
BC116408.1 | - | 5, 6 | 1176 | |||
BC128493.1 | - | 5, 6 | 1173 | |||
BC144792.1 | - | 7, 8 | 1486 | |||
BC145739.1 | - | 7, 8 | 1486 | |||
BC152360.1 | - | 5, 6 | 1173 | |||
BC152361.1 | - | 5, 6 | 1161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
