siRNA Details
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 18 | 5113 | ||
GenBank mRNA | AK012129.1 | - | 1 | 87 | ||
AK044193.1 | - | 5 | 940 | |||
AK087819.1 | - | 5 | 1098 | |||
AK144537.1 | - | 3 | 358 | |||
AK172968.1 | - | 11 | 1856 | |||
BC095969.1 | - | 8 | 1646 | |||
BC116408.1 | - | 3 | 706 | |||
BC128493.1 | - | 3 | 706 | |||
BC144792.1 | - | 5 | 1019 | |||
BC145739.1 | - | 5 | 1019 | |||
BC152360.1 | - | 3 | 706 | |||
BC152361.1 | - | 3 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
