siRNA Details
Gene Symbol: Spag17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028892.4 | NP_083168.3 | 11 | 1500 | ||
GenBank mRNA | AK016477.1 | - | 11 | 1486 | ||
AK029691.1 | - | 11 | 1501 | |||
AK029947.1 | - | 11 | 1496 | |||
AY555275.1 | - | 11 | 1486 | |||
AY792594.1 | - | 11 | 1486 |
