siRNA Details
Gene Symbol: 4930452B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028934.3 | NP_083210.3 | 5, 6 | 684 | ||
GenBank mRNA | AK016701.1 | - | 5, 6 | 633 | ||
AK019627.1 | - | 5, 6 | 641 | |||
AK133893.1 | - | 5, 6 | 686 | |||
BC064468.1 | - | 5, 6 | 634 | 6333742 |
