siRNA Details
Gene Symbol: 4933427D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028963.2 | NP_083239.1 | 3 | 884 | ||
GenBank mRNA | AK016946.1 | - | 3 | 773 | ||
AK032379.1 | - | 3 | 778 | |||
AK041396.1 | - | 3 | 765 | |||
AK043241.1 | - | 3 | 943 | |||
AK173018.1 | - | 3 | 753 | |||
BC043106.1 | - | 3 | 884 | 6406803 | ||
BC059269.1 | - | 3 | 970 |
