siRNA Details
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290148.1 | NP_001277077.1 | 11, 12 | 1073 | ||
NM_028964.4 | NP_083240.2 | 18, 19 | 2122 | |||
GenBank mRNA | AF399755.1 | - | 11, 12 | 1028 | ||
AK017102.1 | - | 11, 12 | 1074 |
