siRNA Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 4 | 274 | ||
GenBank mRNA | AK015776.1 | - | 4 | 253 | ||
AK030743.1 | - | 4 | 309 | |||
AK150733.1 | - | 4 | 272 | |||
AK151141.1 | - | 4 | 272 | |||
AK151382.1 | - | 4 | 272 | |||
AK161646.1 | - | 4 | 295 | |||
AK168966.1 | - | 4 | 298 | |||
AK169324.1 | - | 4 | 318 | |||
BC009141.1 | - | 4 | 274 | 3598647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
