siRNA Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 5 | 387 | ||
GenBank mRNA | AK015776.1 | - | 5 | 366 | ||
AK030743.1 | - | 5 | 422 | |||
AK150733.1 | - | 5 | 385 | |||
AK151141.1 | - | 5 | 385 | |||
AK151382.1 | - | 5 | 385 | |||
AK161646.1 | - | 5 | 408 | |||
AK168966.1 | - | 5 | 411 | |||
AK169324.1 | - | 5 | 431 | |||
BC009141.1 | - | 5 | 387 | 3598647 | ||
BC092058.1 | - | 5 | 419 | 4017388 |

More Information
Other Species Targeted

Gene Ontology Categories:
