siRNA Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 6, 7 | 513 | ||
GenBank mRNA | AK030743.1 | - | 6, 7 | 548 | ||
AK150733.1 | - | 6, 7 | 511 | |||
AK151141.1 | - | 6, 7 | 511 | |||
AK151382.1 | - | 6, 7 | 511 | |||
AK161646.1 | - | 6, 7 | 534 | |||
AK168966.1 | - | 6, 7 | 537 | |||
AK169324.1 | - | 6, 7 | 557 | |||
BC009141.1 | - | 6, 7 | 513 | 3598647 | ||
BC092058.1 | - | 6, 7 | 545 | 4017388 |

More Information
Other Species Targeted

Gene Ontology Categories:
