siRNA Details
Gene Symbol: 1700009J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005796.1 | - | 1 | 630 | ||
AK006749.1 | - | 1 | 614 | |||
AK015973.1 | - | 1 | 624 | |||
BC048578.1 | - | 1 | 474 | |||
BC061019.1 | - | 1 | 629 |
