siRNA Details
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027261.3 | NP_081537.1 | 6 | 1230 | ||
NM_029248.2 | NP_083524.2 | 7 | 1017 | |||
GenBank mRNA | AK016109.1 | - | 7 | 971 | ||
AK051978.1 | - | 1 | 1615 | |||
BC056964.1 | - | 5 | 1168 | |||
BC117068.1 | - | 6 | 949 | 40126387 | ||
BC117070.1 | - | 6 | 949 | 40126389 | ||
CB523122.1 | - | 1 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
