siRNA Details
Gene Symbol: Cfap126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081275.1 | NP_001074744.1 | 6 | 853 | ||
GenBank mRNA | AK005804.1 | - | 6 | 855 | ||
BC061017.1 | - | 5 | 579 | 6774925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
