siRNA Details
Gene Symbol: 9130401M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029418.4 | NP_083694.1 | 5 | 918 | ||
GenBank mRNA | AK018655.1 | - | 5 | 842 | ||
AK047927.1 | - | 5 | 885 | |||
AK089715.1 | - | 842 | ||||
AK167012.1 | - | 5 | 940 | |||
AY053393.1 | - | 5 | 941 | |||
BC019392.1 | - | 4 | 1224 | |||
BC039940.1 | - | 4 | 1247 | |||
BC050895.1 | - | 5 | 918 | 5714794 |
