siRNA Details
Gene Symbol: 9130401M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029418.4 | NP_083694.1 | 6 | 1203 | ||
GenBank mRNA | AK018655.1 | - | 6 | 1127 | ||
AK047927.1 | - | 6 | 1170 | |||
AK089715.1 | - | 1127 | ||||
AK167012.1 | - | 6 | 1225 | |||
AK207239.1 | - | 1 | 15 | |||
AK216091.1 | - | 1 | 15 | |||
AY053393.1 | - | 6 | 1226 | |||
BC022163.1 | - | 1 | 1907 | |||
BC050895.1 | - | 6 | 1203 | 5714794 |
