siRNA Details
Gene Symbol: UQCRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199975.2 | NP_001186904.1 | 5 | 853 | ||
NM_001254752.1 | NP_001241681.1 | 5 | 876 | |||
NM_006294.4 | NP_006285.1 | 4 | 739 | |||
NR_045639.1 | 5 | 1029 | ||||
GenBank mRNA | AK295007.1 | - | 5 | 941 | ||
AK307085.1 | - | 5 | 762 | |||
BC005230.1 | - | 4 | 688 | 3961169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
