siRNA Details
Gene Symbol: Cdyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029441.3 | NP_083717.1 | 3 | 903 | ||
GenBank mRNA | AK006509.1 | - | 2 | 198 | ||
AK015452.1 | - | 3 | 757 | |||
AK142084.1 | - | 3 | 793 | |||
AY273799.1 | - | 3 | 752 | |||
BC058956.1 | - | 3 | 903 | 5690642 |

More Information
Other Species Targeted

Gene Ontology Categories:
