siRNA Details
Gene Symbol: Fam118a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133750.4 | NP_598511.1 | 9 | 1352 | ||
GenBank mRNA | AK143690.1 | - | 9 | 1325 | ||
BC016425.1 | - | 9 | 1210 | 4487028 | ||
BC058794.1 | - | 9 | 1413 | 6486049 | ||
BC066174.1 | - | 9 | 1268 | 30109713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
