siRNA Details
Gene Symbol: Fam118a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133750.4 | NP_598511.1 | 9 | 2014 | ||
GenBank mRNA | AK014192.1 | - | 1 | 396 | ||
AK143690.1 | - | 9 | 1988 | |||
BC016425.1 | - | 9 | 1869 | 4487028 | ||
BC058794.1 | - | 9 | 2070 | 6486049 | ||
BC066174.1 | - | 9 | 1931 | 30109713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
