siRNA Details
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199177.1 | NP_001186106.1 | 6 | 851 | ||
NM_133752.3 | NP_598513.1 | 5 | 797 | |||
GenBank mRNA | AB044138.1 | - | 5 | 789 | ||
AK004715.1 | - | 6 | 807 | |||
AK029157.1 | - | 5 | 755 | |||
AK030835.1 | - | 6 | 805 | |||
AK038446.1 | - | 5 | 744 | |||
AK044657.1 | - | 5 | 730 | |||
AK050383.1 | - | 5 | 737 | |||
AK053861.1 | - | 5 | 755 | |||
AK122311.1 | - | 5 | 725 | |||
AK145620.1 | - | 5 | 749 | |||
BC052704.1 | - | 5 | 739 | |||
BC138665.1 | - | 5 | 678 | 8861686 | ||
BC145959.1 | - | 5 | 678 | 40131262 |
