siRNA Details
Gene Symbol: Pitpnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145823.2 | NP_665822.1 | 2 | 970 | ||
GenBank mRNA | AB077281.1 | - | 2 | 951 | ||
AB077282.1 | - | 2 | 248 | |||
AK003844.1 | - | 1 | 218 | |||
AK021206.1 | - | 2 | 183 | |||
AK153625.1 | - | 2 | 971 | |||
AK171340.1 | - | 2 | 997 | |||
AK171918.1 | - | 2 | 971 | |||
AK172137.1 | - | 2 | 971 | |||
BC082333.1 | - | 2 | 802 | |||
BC108351.1 | - | 2 | 830 | 6312193 |
