siRNA Details
Gene Symbol: Arhgap15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153820.3 | NP_722542.2 | 9, 10 | 904 | ||
GenBank mRNA | AK036520.1 | - | 9, 10 | 904 | ||
AK040796.1 | - | 10, 11 | 974 | |||
AK170537.1 | - | 9, 10 | 902 | |||
BC024887.1 | - | 9, 10 | 858 | 3156334 | ||
BC034881.1 | - | 9, 10 | 877 | 3468639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
