siRNA Details
Gene Symbol: Perm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172417.3 | NP_766005.2 | 2 | 1353 | ||
GenBank mRNA | AK036660.1 | - | 2 | 1355 | ||
AK040833.1 | - | 2 | 1347 | |||
BC117871.1 | - | 1 | 1128 | 40095536 | ||
BC117872.1 | - | 1 | 1128 | 40095540 | ||
KF150176.1 | - | 1 | 1020 |
