siRNA Details
Gene Symbol: Dmkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166173.1 | NP_001159645.1 | 4 | 804 | ||
NM_001166174.1 | NP_001159646.1 | 4 | 804 | |||
NM_172899.4 | NP_766487.3 | 4 | 804 | |||
GenBank mRNA | AK081753.1 | - | 4 | 806 | ||
AY444557.1 | - | 4 | 761 | |||
AY622963.1 | - | 4 | 636 | |||
BC064724.1 | - | 4 | 797 | 30435974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
