siRNA Details
Gene Symbol: 2900052N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038437.1 | - | 3 | 1183 | ||
AK049847.1 | - | 2 | 1263 | |||
AK140089.1 | - | 2 | 1235 | |||
BC054400.1 | - | 2 | 1239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
