siRNA Details
Gene Symbol: C130026I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175219.3 | NP_780428.1 | 4 | 893 | ||
GenBank mRNA | AK047983.1 | - | 4 | 871 | ||
BC007193.1 | - | 484 | 3593825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
