siRNA Details
Gene Symbol: NEBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173484.1 | NP_001166955.1 | 7 | 3560 | ||
NM_006393.2 | NP_006384.1 | 28 | 5837 | |||
NM_213569.2 | NP_998734.1 | 7 | 3562 | |||
XM_005252342.4 | XP_005252399.1 | 27 | 5839 | |||
XM_005252343.4 | XP_005252400.1 | 26 | 5698 | |||
XM_005252344.4 | XP_005252401.1 | 27 | 5742 | |||
XM_011519291.2 | XP_011517593.1 | 30 | 5833 | |||
XM_017015467.1 | XP_016870956.1 | 30 | 5949 | |||
XM_017015468.1 | XP_016870957.1 | 31 | 5717 | |||
GenBank mRNA | AB209311.1 | - | 7 | 3434 | ||
Y16241.1 | - | 28 | 5833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
