siRNA Details
Gene Symbol: NEDD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142393.1 | NP_001135865.1 | 6 | 1189 | ||
NM_001271033.1 | NP_001257962.1 | 4 | 547 | |||
NM_006403.3 | NP_006394.1 | 5 | 994 | |||
NR_073131.1 | 8 | 1434 | ||||
GenBank mRNA | AK292682.1 | - | 6 | 1189 | ||
BC040207.1 | - | 5 | 980 | 4820745 | ||
BP262776.1 | - | 4 | 528 | |||
BX648041.1 | - | 8 | 1401 | |||
L43821.1 | - | 5 | 990 | |||
U64317.1 | - | 5 | 965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
