siRNA Details
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289006.1 | NP_001275935.1 | 6 | 2176 | ||
NM_006405.6 | NP_006396.2 | 6 | 2382 | |||
GenBank mRNA | AK292110.1 | - | 6 | 2071 | ||
BC010856.2 | - | 5 | 1991 | 3899246 | ||
BX161390.1 | - | 6 | 2302 | |||
U94831.1 | - | 5 | 1988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
