siRNA Details
Gene Symbol: Ttc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183106.2 | NP_898929.2 | 18 | 2618 | ||
GenBank mRNA | AK018641.1 | - | 2 | 258 | ||
BC061503.1 | - | 8 | 1191 | |||
BC095993.1 | - | 3 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
