siRNA Details
Gene Symbol: Ttc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183106.2 | NP_898929.2 | 25 | 4183 | ||
GenBank mRNA | AK018641.1 | - | 9 | 1823 | ||
BC061503.1 | - | 15 | 2756 | |||
BC095993.1 | - | 10 | 2048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
