siRNA Details
Gene Symbol: 4930474M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015576.1 | - | 3 | 479 | ||
BC089489.1 | - | 3 | 454 | 6741101 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183107.1 | - | 479 | Chr. - - [-] on Build |
